missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzenecarboximidamide hydrochloride, ≥95%, Thermo Scientific™

£32.20 - £245.00

Chemical Identifiers

CAS 97603-94-6
Molecular Formula C9H7ClF6N2
Molecular Weight (g/mol) 292.609
MDL Number MFCD00276604
InChI Key GOCQSQHVIVLYOG-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzamidine hydrochloride, 3,5-bis trifluoromethyl benzenecarboximidamide hydrochloride, acmc-20andy, 3,5-bis trifluoromethyl benzimidamide hydrochloride, 3,5-bis trifluoromethyl benzenecarboxamidine, chloride, 3,5-bis trifluoromethyl benzene-1-carboximidamide-hydrogen chloride 1/1
PubChem CID 13590400
IUPAC Name 3,5-bis(trifluoromethyl)benzenecarboximidamide;hydrochloride
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=N)N.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10136802
View Documents
Thermo Scientific Maybridge
RF00749DA
1 g Amber Glass Bottle
£32.20
1g
Please to purchase this item. Need a web account? Register with us today!
10320783
View Documents
Thermo Scientific Maybridge
10 g Amber Glass Bottle
£245.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

97603-94-6
292.609
GOCQSQHVIVLYOG-UHFFFAOYSA-N
13590400
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=N)N.Cl
C9H7ClF6N2
MFCD00276604
3,5-bis trifluoromethyl benzamidine hydrochloride, 3,5-bis trifluoromethyl benzenecarboximidamide hydrochloride, acmc-20andy, 3,5-bis trifluoromethyl benzimidamide hydrochloride, 3,5-bis trifluoromethyl benzenecarboxamidine, chloride, 3,5-bis trifluoromethyl benzene-1-carboximidamide-hydrogen chloride 1/1
3,5-bis(trifluoromethyl)benzenecarboximidamide;hydrochloride

Specifications

97603-94-6
95%
MFCD00276604
GOCQSQHVIVLYOG-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)benzenecarboximidamide;hydrochloride
13590400
95%
95.0
C9H7ClF6N2
3,5-bis trifluoromethyl benzamidine hydrochloride, 3,5-bis trifluoromethyl benzenecarboximidamide hydrochloride, acmc-20andy, 3,5-bis trifluoromethyl benzimidamide hydrochloride, 3,5-bis trifluoromethyl benzenecarboxamidine, chloride, 3,5-bis trifluoromethyl benzene-1-carboximidamide-hydrogen chloride 1/1
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=N)N.Cl
292.609
292.61
3, 5-Bis(trifluoromethyl)benzamidine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications