missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11414053
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39234-86-1
Molecular Formula C8H3ClF6O2S
Molecular Weight (g/mol) 312.61
MDL Number MFCD00014725
InChI Key BTRCVKADYDVSLI-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724
PubChem CID 520945
IUPAC Name 3,5-bis(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F

Specifications

Melting Point 34°C to 38°C
Quantity 25 g
UN Number UN3261
Beilstein 3621218
Sensitivity Moisture sensitive
Formula Weight 312.61
Percent Purity 97%
Chemical Name or Material 3,5-Bis (trifluoromethyl)benzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'