missing translation for 'onlineSavingsMsg'
Learn More
3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11494043

3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11494043
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11494043

Brand: Thermo Scientific Alfa Aesar A10300.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39234-86-1
Molecular Formula C8H3ClF6O2S
Molecular Weight (g/mol) 312.61
MDL Number MFCD00014725
InChI Key BTRCVKADYDVSLI-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724
PubChem CID 520945
IUPAC Name 3,5-bis(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F

Specifications

Melting Point 34°C to 38°C
Quantity 1 g
UN Number UN3261
Beilstein 3621218
Sensitivity Moisture sensitive
Formula Weight 312.61
Percent Purity 97%
Chemical Name or Material 3,5-Bis (trifluoromethyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.