missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store away from oxidizing agents.
3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
Density | 1.42 |
Flash Point | >110°C (230°F) |
Refractive Index | 1.4234 |
Quantity | 1 g |
UN Number | UN3276 |
Beilstein | 6810362 |
Solubility Information | Difficult to mix. |
Formula Weight | 253.15 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Bis(trifluoromethyl)phenylacetonitrile |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction