missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid, 98%, Thermo Scientific Chemicals

Product Code. 11434347
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20170-32-5
MDL Number MFCD00017519
Synonym 3-3,5-di-tert-butyl-4-hydroxyphenyl propionic acid, 3-3,5-di-tert-butyl-4-hydroxyphenyl propanoic acid, fenozan, 3,5-di-tert-butyl-4-hydroxyphenylpropionic acid, benzenepropanoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-x6g09g700f, 3,5-di-tert-butyl-4-hydroxyhydocinnamic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxybenzenepropanoic acid, 3-3,5-ditertbutyl-4-hydroxyphenyl propionic acid, 3-3,5-bis tert-butyl-4-hydroxyphenyl propanoic acid
PubChem CID 88389
IUPAC Name 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoic acid

Specifications

Melting Point 172°C to 175°C
Molecular Formula C17H26O3
Quantity 5 g
Solubility Information Slightly soluble in water.
InChI Key WPMYUUITDBHVQZ-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CCC(=O)O
Molecular Weight (g/mol) 278.392
Formula Weight 278.4
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'