missing translation for 'onlineSavingsMsg'
Learn More
3,5-Diamino-1,2,4-triazole, 98%, Thermo Scientific Chemicals Product Code.: 10489210

3,5-Diamino-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Product Code. 10489210
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10489210

Brand: Thermo Scientific Acros 112440250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-77-2
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.09
MDL Number MFCD00005233
InChI Key PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Synonym guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
PubChem CID 15078
ChEBI CHEBI:75425
IUPAC Name 1H-1,2,4-triazole-3,5-diamine
SMILES C1(=NC(=NN1)N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 204°C to 208°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 26, 193
Formula Weight 99.09
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3,5-Diamino-1,2,4-triazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468967 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number112440250,112441000
Lot NumberA0462815 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number112440250,112441000
Lot NumberA0430885 Certificate TypeCertificate of Analysis Date04/12/2023 Catalog Number112440050,112440250,112441000
Lot NumberA0418022 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number112440050,112440250,112441000
Lot NumberA0445130 Certificate TypeCertificate of Analysis Date17/08/2022 Catalog Number112440050,112440250,112441000
Product Identifier
  • 3,5-Diamino-1,2,4-triazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Diamino-1,2,4-triazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.