missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromo-1H-1,2,4-triazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 226.86 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Dibromo-1H-1,2,4-triazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'