Learn More
3,5-Dibromobenzaldehyde, 98+%, Thermo Scientific Chemicals
£27.30 - £327.00
Chemical Identifiers
CAS | 56990-02-4 |
---|---|
Molecular Formula | C7H4Br2O |
Molecular Weight (g/mol) | 263.91 |
MDL Number | MFCD00156887 |
InChI Key | ZLDMZIXUGCGKMB-UHFFFAOYSA-N |
Synonym | 3,5-dibromo-benzaldehyde, benzaldehyde, 3,5-dibromo, pubchem3069, acmc-209lvd, ksc493o2p, 3,5-bis bromanyl benzaldehyde, paragos 530307, 3,5-dibromo-phenyl-methanone |
PubChem CID | 622077 |
IUPAC Name | 3,5-dibromobenzaldehyde |
SMILES | C1=C(C=C(C=C1Br)Br)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10298373
|
Thermo Scientific Acros
393150010 |
1 g | Glass bottle |
£27.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10005353
|
Thermo Scientific Acros
393150050 |
5 g | Glass bottle |
£88.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10678804
|
Thermo Scientific Acros
393150250 |
25 g | Glass bottle |
£327.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56990-02-4 | |
263.91 | |
ZLDMZIXUGCGKMB-UHFFFAOYSA-N | |
622077 | |
C1=C(C=C(C=C1Br)Br)C=O |
C7H4Br2O | |
MFCD00156887 | |
3,5-dibromo-benzaldehyde, benzaldehyde, 3,5-dibromo, pubchem3069, acmc-209lvd, ksc493o2p, 3,5-bis bromanyl benzaldehyde, paragos 530307, 3,5-dibromo-phenyl-methanone | |
3,5-dibromobenzaldehyde |
Specifications
56990-02-4 | |
100.0 | |
White to Beige | |
98% min. (GC) | |
Br2C6H3C(=O)H | |
3,5-dibromo-benzaldehyde, benzaldehyde, 3,5-dibromo, pubchem3069, acmc-209lvd, ksc493o2p, 3,5-bis bromanyl benzaldehyde, paragos 530307, 3,5-dibromo-phenyl-methanone | |
ZLDMZIXUGCGKMB-UHFFFAOYSA-N | |
3,5-dibromobenzaldehyde | |
622077 | |
98+% | |
3, 5-Dibromobenzaldehyde |
98.0 | |
86°C to 90°C | |
Authentic | |
C7H4Br2O | |
MFCD00156887 | |
Solubility in water: insoluble. Other solubilities: soluble in dmso,ethyl acetate,methanol | |
C1=C(C=C(C=C1Br)Br)C=O | |
263.91 | |
263.91 | |
Crystals or Powder |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
Recommended Storage : 4°C
- 3,5-Dibromobenzaldehyde
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only