missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-dibromobenzamide, Thermo Scientific™
£25.30 - £286.00
Chemical Identifiers
CAS | 175205-85-3 |
---|---|
Molecular Formula | C7H5Br2NO |
Molecular Weight (g/mol) | 278.931 |
InChI Key | IOFGMNBDIDEBIQ-UHFFFAOYSA-N |
Synonym | 3,5-dibromo-benzoic acid amide, acmc-20andi, 3,5-dibromo-benzamide, maybridge1_007750, 3,5-bis bromanyl benzamide, 3,5-dibromobenzamide |
PubChem CID | 2735937 |
IUPAC Name | 3,5-dibromobenzamide |
SMILES | C1=C(C=C(C=C1Br)Br)C(=O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10032504
|
Thermo Scientific Maybridge
SEW04141DA |
1 g |
£25.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10452464
|
Thermo Scientific Maybridge
SEW04141EA |
10 g |
£145.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10683445
|
Thermo Scientific Maybridge
SEW04141EB |
25 g |
£286.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
175205-85-3 | |
278.931 | |
3,5-dibromo-benzoic acid amide, acmc-20andi, 3,5-dibromo-benzamide, maybridge1_007750, 3,5-bis bromanyl benzamide, 3,5-dibromobenzamide | |
3,5-dibromobenzamide |
C7H5Br2NO | |
IOFGMNBDIDEBIQ-UHFFFAOYSA-N | |
2735937 | |
C1=C(C=C(C=C1Br)Br)C(=O)N |
Specifications
175205-85-3 | |
C7H5Br2NO | |
IOFGMNBDIDEBIQ-UHFFFAOYSA-N | |
3,5-dibromobenzamide | |
2735937 | |
97% |
97% | |
3,5-dibromo-benzoic acid amide, acmc-20andi, 3,5-dibromo-benzamide, maybridge1_007750, 3,5-bis bromanyl benzamide, 3,5-dibromobenzamide | |
C1=C(C=C(C=C1Br)Br)C(=O)N | |
278.931 | |
278.93 | |
3, 5-Dibromobenzamide |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title