missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11476706
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117695-55-3
Molecular Formula C6H5BBr2O2
Molecular Weight (g/mol) 279.722
MDL Number MFCD01075725
InChI Key WQBLCGDZYFKINX-UHFFFAOYSA-N
Synonym 3,5-dibromobenzeneboronic acid, 3,5-dibromophenyl boronic acid, boronic acid, 3,5-dibromophenyl, pubchem14108, acmc-2099ur, amtb596, 3,5-dibromobenzeneboronicacid, ksc493i3d, 3,5-dibromobenzene boronic acid
PubChem CID 2734689
IUPAC Name (3,5-dibromophenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)Br)Br)(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 6645285
Formula Weight 279.73
Percent Purity 97%
Chemical Name or Material 3,5-Dibromobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'