missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromobenzoic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3,5-Dibromobenzoic acid may be used for the syntheses (+)-menthyl 3,5-dibromobenzoate, di-tert-butyl 4-[2-(tert-butoxycarbonyl)ethyl]-4-(3,5-dibromobenzamido)heptanedioate2, (L)-methyl 2-(3,5-dibromobenzamido)-3-phenylpropanoate.
Solubility
Very faint turbidity in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
3,5-Dibromobenzoic acid may be used for the syntheses (+)-menthyl 3,5-dibromobenzoate, di-tert-butyl 4-[2-(tert-butoxycarbonyl)ethyl]-4-(3,5-dibromobenzamido)heptanedioate2, (L)-methyl 2-(3,5-dibromobenzamido)-3-phenylpropanoate.
Solubility
Very faint turbidity in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 216°C to 224°C |
Quantity | 1 g |
Beilstein | 1940691 |
Solubility Information | Very faint turbidity in methanol. |
Formula Weight | 279.93 |
Percent Purity | 98% |
Chemical Name or Material | 3,5-Dibromobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'