missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromopyridin-2-amine, 97%, Thermo Scientific™

Product Code. 10555134
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35486-42-1
Molecular Formula C5H4Br2N2
Molecular Weight (g/mol) 251.909
MDL Number MFCD00038041
InChI Key WJMJWMSWJSACSN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromopyridine, 3,5-dibromo-2-pyridylamine, 2-pyridinamine, 3,5-dibromo, 3,5-dibromo-2-pyridinamine, 3,5-dibromo-pyridin-2-ylamine, 3,5-dibromo-2-pyridinylamine, 2-amino-3,5-dibromo pyridine, 3,5-dibrompyridin-2-amin, pubchem1130, maybridge1_001693
PubChem CID 98851
IUPAC Name 3,5-dibromopyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula Br2(C5H2N)NH2
Quantity 10 g
Formula Weight 251.91
Percent Purity 97%
Chemical Name or Material 3, 5-Dibromopyridin-2-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.