missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-pyridone-1-acetic acid, 98%, Thermo Scientific™

Product Code. 11457146
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dichloro-4-pyridone-1-acetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56187-37-2
Molecular Formula C7H5Cl2NO3
Molecular Weight (g/mol) 222.021
MDL Number MFCD02180547
InChI Key MFMXEEQESUEMSB-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-pyridone-1-acetic acid, 3,5-dichloro-4-pyridone-n-acetic acid, 2-3,5-dichloro-4-oxopyridin-1 4h-yl acetic acid, 1 4h-pyridineacetic acid, 3,5-dichloro-4-oxo, 3,5-dichloro-4-oxo-4h-pyridine-1-acetic acid, 3,5-dichloro-4-oxopyridin-1-yl acetic acid, 3,5-dichloro-4-pyridine-n-acetic acid, 2-3,5-dichloro-4-oxohydropyridyl acetic acid, pubchem11174, ksc272s7b
PubChem CID 92052
IUPAC Name 2-(3,5-dichloro-4-oxopyridin-1-yl)acetic acid
SMILES C1=C(C(=O)C(=CN1CC(=O)O)Cl)Cl

Specifications

Melting Point 239°C to 241°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 222.03
Percent Purity 98%
Chemical Name or Material 3,5-Dichloro-4-pyridone-1-acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'