missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzaldehyde 97%, ACROS Organics™

Catalog Number 10772771
Click to view available options
1g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 10203-08-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 61°C to 66°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H4Cl2O
Linear Formula Cl2C6H3CHO
MDL Number MFCD00003352
Quantity 1 g
Synonym benzaldehyde, 3,5-dichloro, 3,5-dichloro-benzaldehyde, unii-m74yu7se79, rarechem aq a2 0002, pubchem3068, 3,5-dichlorobezaldehyde, 3,5-dichlorobenaldehyde, 3?5-dichlorobenzaldehyde, benzaldehyde,5-dichloro, acmc-1bxq6
InChI Key CASRSOJWLARCRX-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1Cl)Cl)C=O
IUPAC Name 3,5-dichlorobenzaldehyde
Molecular Weight (g/mol) 175.008
PubChem CID 35746
Formula Weight 175.01
Percent Purity ≥96.0%
Physical Form Crystalline Mass or Fluffy Powder
Chemical Name or Material 3, 5-Dichlorobenzaldehyde, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'