missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichlorobenzene-1-carboximidamide hydrochloride, Tech., Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Linear Formula | Cl2C6H3C(=NH)NH2·HCl |
Quantity | 1 g |
IUPAC Name | 3,5-dichlorobenzenecarboximidamide;hydrochloride |
Formula Weight | 225.51 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 3, 5-Dichlorobenzene-1-carboximidamide hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'