Learn More
3,5-Dichlorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
£21.30 - £205.00
Chemical Identifiers
CAS | 705-21-5 |
---|---|
Molecular Formula | C6H3Cl3O2S |
Molecular Weight (g/mol) | 245.50 |
MDL Number | MFCD00051698 |
InChI Key | RJSQINMKOSOUGT-UHFFFAOYSA-N |
Synonym | 3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t |
PubChem CID | 522388 |
IUPAC Name | 3,5-dichlorobenzenesulfonyl chloride |
SMILES | ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494453
|
Thermo Scientific Alfa Aesar
A11003.03 |
1 g |
£21.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404463
|
Thermo Scientific Alfa Aesar
A11003.06 |
5 g |
£62.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414463
|
Thermo Scientific Alfa Aesar
A11003.14 |
25 g |
£205.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
705-21-5 | |
245.50 | |
RJSQINMKOSOUGT-UHFFFAOYSA-N | |
522388 | |
ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O |
C6H3Cl3O2S | |
MFCD00051698 | |
3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t | |
3,5-dichlorobenzenesulfonyl chloride |
Specifications
705-21-5 | |
158°C to 160°C (15 mmHg) | |
C6H3Cl3O2S | |
UN3261 | |
Moisture sensitive | |
RJSQINMKOSOUGT-UHFFFAOYSA-N | |
3,5-dichlorobenzenesulfonyl chloride | |
522388 | |
97% |
30°C to 33°C | |
>110°C (230°F) | |
MFCD00051698 | |
1528657 | |
3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t | |
ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O | |
245.50 | |
245.51 | |
3,5-Dichlorobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only