missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzoic acid, 99%

Product Code. 11452168
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11452168

Brand: Thermo Scientific Alfa Aesar A15168.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-36-5 | C7H4Cl2O2 | 191.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-36-5
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00002494
InChI Key CXKCZFDUOYMOOP-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dichloro, 3,5-dichloro-benzoic acid, unii-7baa7r70gn, 7baa7r70gn, ccris 8611, acmc-209ktc, 3,5-dichlorobenzoicacid, 3,5-dichlorobenzic acid, 3 5-dichlorobenzoic acid, dsstox_cid_4981
PubChem CID 5811
IUPAC Name 3,5-dichlorobenzoic acid
SMILES C1=C(C=C(C=C1Cl)Cl)C(=O)O

Specifications

Melting Point 185°C to 187°C
Quantity 100 g
Beilstein 2044776
Formula Weight 191.01
Percent Purity 99%
Chemical Name or Material 3,5-Dichlorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.