Learn More
3,5-Dichlorophenyl isocyanate, 96%, Thermo Scientific Chemicals
£22.10 - £184.00
Chemical Identifiers
CAS | 34893-92-0 |
---|---|
Molecular Formula | C7H3Cl2NO |
Molecular Weight (g/mol) | 188.01 |
MDL Number | MFCD00013859 |
InChI Key | XEFUJGURFLOFAN-UHFFFAOYSA-N |
Synonym | 3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate |
PubChem CID | 94460 |
IUPAC Name | 1,3-dichloro-5-isocyanatobenzene |
SMILES | C1=C(C=C(C=C1Cl)Cl)N=C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10221053
|
Thermo Scientific Acros
314140010 |
1 g | Glass bottle |
£22.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10033570
|
Thermo Scientific Acros
314140050 |
5 g | Glass bottle |
£55.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10599453
|
Thermo Scientific Acros
314140250 |
25 g | Glass bottle |
£184.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34893-92-0 | |
188.01 | |
XEFUJGURFLOFAN-UHFFFAOYSA-N | |
94460 | |
C1=C(C=C(C=C1Cl)Cl)N=C=O |
C7H3Cl2NO | |
MFCD00013859 | |
3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate | |
1,3-dichloro-5-isocyanatobenzene |
Specifications
34893-92-0 | |
100.0 | |
White to Yellow | |
243.0°C | |
Authentic | |
C7H3Cl2NO | |
MFCD00013859 | |
3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate | |
XEFUJGURFLOFAN-UHFFFAOYSA-N | |
1,3-dichloro-5-isocyanatobenzene | |
94460 | |
96% | |
3, 5-Dichlorophenyl isocyanate |
95.0 | |
29.0°C to 35.0°C | |
1.3800g/mL | |
110°C | |
96% | |
Cl2C6H3NCO | |
1.38 | |
Solubility in water: decomposes | |
C1=C(C=C(C=C1Cl)Cl)N=C=O | |
188.01 | |
188.01 | |
Low Melting Solid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 252-276-9
RUO – Research Use Only