missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15425315
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 312692-86-7
Density 1.002
Quantity 50 mL
Molecular Formula C6H3Cl2IZn
MDL Number MFCD01319865
UN Number UN3399
Synonym 3,5-dichlorophenylzinc iodide, 3,5-dichlorophenyl zincylium iodide
InChI Key HIWLJUNTNGOHMX-UHFFFAOYSA-M
SMILES C1=[C-]C=C(C=C1Cl)Cl.[Zn+]I
IUPAC Name 1,3-dichlorobenzene-5-ide;iodozinc(1+)
Molecular Weight (g/mol) 338.274
PubChem CID 3341003
Formula Weight 338.28
Chemical Name or Material 3,5-Dichlorophenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Afficher plus Afficher moins
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'