missing translation for 'onlineSavingsMsg'
Learn More
3,5-Difluoropyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 11309176

3,5-Difluoropyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11309176
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11309176

Brand: Thermo Scientific Alfa Aesar L20324.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745784-04-7
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD05664124
InChI Key QKLXAJQKMIWFRC-UHFFFAOYSA-N
Synonym 3,5-difluoropicolinic acid, 3,5-difluoro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3,5-difluoro, 3,5-difluoro-2-pyridinecarboxylicacid, 3,5-difluoropicolinicacid, acmc-209ov6, 2-carboxy-3,5-difluoropyridine, 2-pyridinecarboxylicacid, 3,5-difluoro, 3,5-difluoro-pyridine-2-carboxylic acid, 3 pound not5-difluoropyridine-2-carboxylicacid
PubChem CID 2783262
IUPAC Name 3,5-difluoropyridine-2-carboxylic acid
SMILES OC(=O)C1=C(F)C=C(F)C=N1

Specifications

Melting Point ∽205°C (decomposition)
Quantity 1 g
Formula Weight 159.09
Percent Purity 97%
Chemical Name or Material 3,5-Difluoropyridine-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Difluoropyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.