missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals

£20.20 - £200.00

Chemical Identifiers

CAS 530-59-6
Molecular Formula C11H12O5
Molecular Weight (g/mol) 224.21
InChI Key PCMORTLOPMLEFB-ONEGZZNKSA-N
Synonym sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PubChem CID 637775
ChEBI CHEBI:15714
IUPAC Name (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10428450
View Documents
Thermo Scientific Acros
115550010
1 g Glass bottle
£20.20
1g
Please to purchase this item. Need a web account? Register with us today!
10356130
View Documents
Thermo Scientific
115550050
5 g Glass bottle
£57.89
5g
Please to purchase this item. Need a web account? Register with us today!
11915121
View Documents
Thermo Scientific Acros
115550250
25 g Glass bottle
£200.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

530-59-6
224.21
sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
CHEBI:15714
COC1=CC(=CC(=C1O)OC)C=CC(=O)O
C11H12O5
PCMORTLOPMLEFB-ONEGZZNKSA-N
637775
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid

Specifications

530-59-6
100.0
Beige-Yellow to Yellow
98%
HOC6H2(OCH3)2CH=CHCO2H
sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PCMORTLOPMLEFB-ONEGZZNKSA-N
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
637775
224.21
Crystalline Powder
97.5
203.0°C to 205.0°C
Authentic
C11H12O5
10,508
Solubility in water: insoluble in water. Other solubilities: soluble in methanol,ethanol and acetone
COC1=CC(=CC(=C1O)OC)C=CC(=O)O
224.21
CHEBI:15714
98%
3, 5-Dimethoxy-4-hydroxycinnamic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 208-487-3

Product Identifier
  • 3,5-Dimethoxy-4-hydroxycinnamic acid, predominantly trans isomer

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only