missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzoyl chloride, 98%, Thermo Scientific™

Product Code. 11497786
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dimethoxybenzoyl chloride was used to study the mechanism and kinetics of solvolysis of 3,4- and 3,5-dimethoxybenzoyl chlorides in various binary solvents. 3,5-Dimethoxybenzoyl chloride undergoes addition reaction with 4,4-dimethyl-2-pentyne in presence of AlCl3 via 1,2- methyl shift. It is used as a chemical and organic intermediate.

Solubility
It hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17213-57-9
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.62
MDL Number MFCD00000676
InChI Key FTHPLWDYWAKYCY-UHFFFAOYSA-N
PubChem CID 87003
IUPAC Name 3,5-dimethoxybenzoyl chloride
SMILES COC1=CC(=CC(OC)=C1)C(Cl)=O

Specifications

Melting Point 42°C to 44°C
Boiling Point 152°C to 154°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 511839
Sensitivity Moisture sensitive
Synonym benzoyl chloride, 3,5-dimethoxy, 3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-benzoyl chloride, acmc-1c560, benzoyl chloride,3,5-dimethoxy, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride
Solubility Information It hydrolyzes in water.
Formula Weight 200.62
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxybenzoyl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'