missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethoxyphenol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.
Solubility
Soluble in methanol, ethanol, benzene, water (slightly), and chloroform.
Notes
Air and light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, light.
3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.
Solubility
Soluble in methanol, ethanol, benzene, water (slightly), and chloroform.
Notes
Air and light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, light.
Specifications
Specifications
Melting Point | 45°C to 47°C |
Boiling Point | 172°C to 175°C (17 mmHg) |
Flash Point | 78°C (173°F) |
Quantity | 10 g |
Beilstein | 1239156 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in methanol,ethanol,benzene,water (slightly),and chloroform. |
Formula Weight | 154.16 |
Percent Purity | 98% |
Chemical Name or Material | 3,5-Dimethoxyphenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'