missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 10496082
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
61226-19-5
C12H11N2O2
215.23
MFCD02656670
LPYTYYLNGJGJGW-UHFFFAOYSA-M
3,5-dimethyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carboxylicacid, 4-carboxy-3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid,3,5-dimethyl-1-phenyl, 1h-pyrazole-4-carboxylicacid, 3,5-dimethyl-1-phenyl, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbox ylic acid
612275
CC1=NN(C(C)=C1C([O-])=O)C1=CC=CC=C1
This item is not returnable. View return policy
61226-19-5
C12H11N2O2
215.23
MFCD02656670
LPYTYYLNGJGJGW-UHFFFAOYSA-M
3,5-dimethyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carboxylicacid, 4-carboxy-3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid,3,5-dimethyl-1-phenyl, 1h-pyrazole-4-carboxylicacid, 3,5-dimethyl-1-phenyl, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbox ylic acid
612275
CC1=NN(C(C)=C1C([O-])=O)C1=CC=CC=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 3,5-dimethyl-1-phenylpyrazole-4-carboxylic acid
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'