missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10374612
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10374612

Brand: Thermo Scientific Maybridge CC01201CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 113808-86-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00159642
InChI Key IOOWDXMXZBYKLR-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl, 1h-pyrazole-4-carboxylicacid, 3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-carboxylicacid, acmc-20a7j9, pyrazole carboxylic acid 1, ksc173i7f, 3,5-dimethylpyrazole-4-carboxylic, 3,5-dimethylpyrazole-4-carboxylicacid, 1h-pyrazole-4-carboxylicacid,3,5-dimethyl
PubChem CID 2776164
IUPAC Name 3,5-dimethyl-1H-pyrazole-4-carboxylic acid
SMILES CC1=C(C(=NN1)C)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-1H-pyrazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.