missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-methoxyphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals

Product Code. 15441586
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

3,5-Dimethyl-4-methoxyphenylmagnesium bromide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyl-4-methoxyphenylmagnesium bromide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Reacts with water.

Note
TRUSTED_SUSTAINABILITY

Specifications

CAS 185416-17-5
Molecular Formula C9H11BrMgO
MDL Number MFCD01319917
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 3,5-dimethyl-4-methoxyphenylmagnesium bromide, kiysunnvtfbqit-uhfffaoysa-m, magnesium,bromo 4-methoxy-3,5-dimethylphenyl, 3,5-dimethyl-4-methoxyphenylmagnesium bromide, 0.5 m in thf, 3,5-dimethyl-4-methoxyphenylmagnesium bromide, 0.5m in 2-methf, 3,5-dimethyl-4-methoxyphenylmagnesium bromide 0.5 m in tetrahydrofuran, grignard reagent
Solubility Information Reacts with water.
InChI Key KIYSUNNVTFBQIT-UHFFFAOYSA-M
SMILES COC1=C(C)C=C([Mg]Br)C=C1C
IUPAC Name magnesium;2-methoxy-1,3-dimethylbenzene-5-ide;bromide
Molecular Weight (g/mol) 239.40
PubChem CID 12914311
Formula Weight 239.39
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 3,5-Dimethyl-4-methoxyphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'