missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylisoxazole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -14°C |
Density | 0.992 |
Boiling Point | 142°C to 144°C |
Flash Point | 31°C (87°F) |
Refractive Index | 1.4415 |
Quantity | 100 g |
UN Number | UN1993 |
Beilstein | 106324 |
Solubility Information | Insoluble in water. |
Formula Weight | 97.12 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'