missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylphenol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).
Solubility
Soluble in ethyl alcohol, carbon tetrachloride, benzene, chloroform, water and ether.
Notes
Incompatible with bases, acid chlorides, acid anhydrides, oxidizing agents, brass and copper.
3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).
Solubility
Soluble in ethyl alcohol, carbon tetrachloride, benzene, chloroform, water and ether.
Notes
Incompatible with bases, acid chlorides, acid anhydrides, oxidizing agents, brass and copper.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 61°C to 65°C |
Density | 1.12 |
Boiling Point | 222°C |
Flash Point | 109°C (228°F) |
Quantity | 500 g |
UN Number | UN2261 |
Beilstein | 774117 |
Merck Index | 14,10082 |
Solubility Information | Soluble in ethyl alcohol,carbon tetrachloride,benzene,chloroform,water and ether. |
Formula Weight | 122.17 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
Signal Word
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'