missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpiperidine, 96%, mixture of cis and trans

Product Code. p-119693 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10114250

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35794-11-7 | C7H16N | 114.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35794-11-7
Molecular Formula C7H16N
Molecular Weight (g/mol) 114.21
MDL Number MFCD00005996,MFCD09832871
InChI Key IDWRJRPUIXRFRX-KNVOCYPGSA-O
Synonym piperidine, 3,5-dimethyl, 3,5-lupetidine, 3,5-dimethylpiperidin, 3,5-dimethylpiperidine, cis + trans, pubchem7709, 3,5-dimethylpiperdine, 3,5 dimethylpiperidine, 3,5-dimethylpiperadine, acmc-1afvq, hexahydro-3,5-lutidine
PubChem CID 118259
IUPAC Name 3,5-dimethylpiperidine
SMILES C[C@H]1C[NH2+]C[C@@H](C)C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 144°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4434 to 1.4464
Quantity 25 g
Specific Gravity 0.85
Formula Weight 113.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3, 5-Dimethylpiperidine, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,5-Dimethylpiperidine, mixture of cis and trans
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.