missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyrazin-2-amine, ≥97%, Thermo Scientific™

Product Code. 10765803
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 91678-81-8
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.159
MDL Number MFCD00107172
InChI Key GZZRWBRYYZGTNW-UHFFFAOYSA-N
Synonym 2-amino-3,5-dimethylpyrazine, 3,5-dimethyl-2-pyrazinamine, 3,5-dimethylpyrazine-2-ylamine, pyrazinamine, 3,5-dimethyl, 2-pyrazinamine, 3,5-dimethyl, 3,5-dimethyl-pyrazinamine, 2-amino-3,5-dimethyl-1,4-diazine, 3,5-dimethylpyrazin-2-ylamine
PubChem CID 2774764
IUPAC Name 3,5-dimethylpyrazin-2-amine
SMILES CC1=CN=C(C(=N1)C)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Quantity 10 g
Formula Weight 123.16
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethylpyrazin-2-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'