missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitro-2-methylbenzeneboronic acid, 96%, Thermo Scientific™

Product Code. 15489367
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24341-76-2
Molecular Formula C7H7BN2O6
Molecular Weight (g/mol) 225.95
MDL Number MFCD00757434
InChI Key ZESXGHPFWNSQQV-UHFFFAOYSA-N
Synonym 3,5-dinitro-2-methylphenylboronic acid, 3,5-dinitro-2-methylphenyl boronic acid, 2-borono-4,6-dinitrotoluene, 2-methyl-3,5-dinitrophenyl boronic acid, 3,5-dinitro-2-methylbenzeneboronic acid, boronicacid, b-2-methyl-3,5-dinitrophenyl, acmc-1ceer, 2-methyl-3,5-dinitro-phenyl boronic acid, boronicacid,b-2-methyl-3,5-dinitrophenyl
PubChem CID 4353496
IUPAC Name (2-methyl-3,5-dinitrophenyl)boronic acid
SMILES CC1=C(C=C(C=C1B(O)O)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 122°C to 128°C
Quantity 5 g
Formula Weight 225.95
Percent Purity 96%
Chemical Name or Material 3,5-Dinitro-2-methylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'