missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoyl Chloride, 99%

Product Code. 10677021 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10677021

Brand: Thermo Scientific Acros 408711000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-33-2 | C7H3ClN2O5 | 230.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-33-2
Molecular Formula C7H3ClN2O5
Molecular Weight (g/mol) 230.56
MDL Number MFCD00007248
InChI Key NNOHXABAQAGKRZ-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
PubChem CID 7432
IUPAC Name 3,5-dinitrobenzoyl chloride
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 67.0°C to 70.0°C
Color Yellow to Beige
Boiling Point 196°C (15.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Beilstein 09, 414
Fieser 01,320
Merck Index 15, 3303
Solubility Information Solubility in water: decomposes
Formula Weight 230.56
Percent Purity 99%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material 3, 5-Dinitrobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.