Learn More
3,5-Dinitrosalicylic acid, 97+%, Thermo Scientific Chemicals
£34.57 - £327.63
Chemical Identifiers
CAS | 609-99-4 |
---|---|
Molecular Formula | C7H4N2O7 |
Molecular Weight (g/mol) | 228.116 |
MDL Number | MFCD00007104 |
InChI Key | LWFUFLREGJMOIZ-UHFFFAOYSA-N |
Synonym | 3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid |
PubChem CID | 11873 |
ChEBI | CHEBI:53648 |
SMILES | C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488826
|
Thermo Scientific Alfa Aesar
A16988.18 |
50 g |
£34.57
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498826
|
Thermo Scientific Alfa Aesar
A16988.30 |
250 g |
£106.53
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478826
|
Thermo Scientific Alfa Aesar
A16988.0B |
1000 g |
£327.63
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.
Solubility
Soluble in water, dimethyl sulfoxide, methanol, ethanol, diethyl ether and benzene.
Notes
Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
Chemical Identifiers
609-99-4 | |
228.116 | |
LWFUFLREGJMOIZ-UHFFFAOYSA-N | |
11873 | |
C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-] |
C7H4N2O7 | |
MFCD00007104 | |
3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid | |
CHEBI:53648 |
Specifications
609-99-4 | |
C7H4N2O7 | |
2220661 | |
Soluble in water,dimethyl sulfoxide,methanol,ethanol,diethyl ether and benzene. | |
C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-] | |
11873 | |
228.12 | |
3,5-Dinitrosalicylic acid |
169°C to 174°C | |
MFCD00007104 | |
3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid | |
LWFUFLREGJMOIZ-UHFFFAOYSA-N | |
228.116 | |
CHEBI:53648 | |
≥97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 210-204-3
missing translation for 'rtecsNumber' : VO2965000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 3,5-Dinitrosalicylic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 3,5-Dinitrosalicylicacid
RUO – Research Use Only