missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Pyrazoledicarboxylic acid monohydrate, 97%
Tous les produits Thermo Scientific Chemicals Produits
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Packaging:
Glass bottle
missing translation for 'unitSize'
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 292°C to 295°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Quantity | 25 g |
| Beilstein | 25, 162 |
| Solubility Information | Solubility in water: insoluble |
| Afficher plus |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu