Learn More
3,6-Dibromocarbazole, 99%, Thermo Scientific Chemicals
£59.70 - £188.00
Chemical Identifiers
CAS | 6825-20-3 |
---|---|
Molecular Formula | C12H7Br2N |
Molecular Weight (g/mol) | 325.003 |
MDL Number | MFCD00004961 |
InChI Key | FIHILUSWISKVSR-UHFFFAOYSA-N |
Synonym | 3,6-dibromocarbazole, 9h-carbazole, 3,6-dibromo, carbazole, 3,6-dibromo, pubchem9951, 3,6-dibromo-carbazole, acmc-209o2m, 3,6,-dibromo-9h-carbazole, 3,6-dibromo-9-h-carbazole, ksc490o8t |
PubChem CID | 274874 |
IUPAC Name | 3,6-dibromo-9H-carbazole |
SMILES | C1=CC2=C(C=C1Br)C3=C(N2)C=CC(=C3)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15412828
|
Thermo Scientific Alfa Aesar
H56360.06 |
5 g |
£59.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15422828
|
Thermo Scientific Alfa Aesar
H56360.14 |
25 g |
£188.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,6-Dibromocarbazole has been used in the preparation of N-(2-hydroxyethyl)-3,6-dibromocarbazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,6-Dibromocarbazole has been used in the preparation of N-(2-hydroxyethyl)-3,6-dibromocarbazole.
Solubility
Soluble in benzene, chloroform
Notes
Incompatible with oxidizing agents. Store at room temperature.
Chemical Identifiers
6825-20-3 | |
325.003 | |
FIHILUSWISKVSR-UHFFFAOYSA-N | |
274874 | |
C1=CC2=C(C=C1Br)C3=C(N2)C=CC(=C3)Br |
C12H7Br2N | |
MFCD00004961 | |
3,6-dibromocarbazole, 9h-carbazole, 3,6-dibromo, carbazole, 3,6-dibromo, pubchem9951, 3,6-dibromo-carbazole, acmc-209o2m, 3,6,-dibromo-9h-carbazole, 3,6-dibromo-9-h-carbazole, ksc490o8t | |
3,6-dibromo-9H-carbazole |
Specifications
6825-20-3 | |
C12H7Br2N | |
3,6-dibromocarbazole, 9h-carbazole, 3,6-dibromo, carbazole, 3,6-dibromo, pubchem9951, 3,6-dibromo-carbazole, acmc-209o2m, 3,6,-dibromo-9h-carbazole, 3,6-dibromo-9-h-carbazole, ksc490o8t | |
FIHILUSWISKVSR-UHFFFAOYSA-N | |
3,6-dibromo-9H-carbazole | |
274874 | |
99% |
204°C to 206°C | |
MFCD00004961 | |
Soluble in benzene,chloroform | |
C1=CC2=C(C=C1Br)C3=C(N2)C=CC(=C3)Br | |
325.003 | |
325 | |
3,6-Dibromocarbazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only