missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',6'-Dichloro-2'-fluoroacetophenone, 97%, Thermo Scientific™
£29.30 - £81.50
Chemical Identifiers
CAS | 916420-72-9 |
---|---|
Molecular Formula | C8H5Cl2FO |
Molecular Weight (g/mol) | 207.025 |
MDL Number | MFCD09025392 |
InChI Key | KIPVBHCZTSPODV-UHFFFAOYSA-N |
Synonym | 3',6'-dichloro-2'-fluoroacetophenone, 1-3,6-dichloro-2-fluorophenyl ethanone, ethanone, 1-3,6-dichloro-2-fluorophenyl, 1-acetyl-3,6-dichloro-2-fluorobenzene |
PubChem CID | 46737559 |
IUPAC Name | 1-(3,6-dichloro-2-fluorophenyl)ethanone |
SMILES | CC(=O)C1=C(C=CC(=C1F)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15454837
|
Thermo Scientific Alfa Aesar
H32149.MD |
250 mg |
£29.30
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444837
|
Thermo Scientific Alfa Aesar
H32149.03 |
1 g |
£81.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
916420-72-9 | |
207.025 | |
KIPVBHCZTSPODV-UHFFFAOYSA-N | |
46737559 | |
CC(=O)C1=C(C=CC(=C1F)Cl)Cl |
C8H5Cl2FO | |
MFCD09025392 | |
3',6'-dichloro-2'-fluoroacetophenone, 1-3,6-dichloro-2-fluorophenyl ethanone, ethanone, 1-3,6-dichloro-2-fluorophenyl, 1-acetyl-3,6-dichloro-2-fluorobenzene | |
1-(3,6-dichloro-2-fluorophenyl)ethanone |
Specifications
916420-72-9 | |
1.5325 | |
3',6'-dichloro-2'-fluoroacetophenone, 1-3,6-dichloro-2-fluorophenyl ethanone, ethanone, 1-3,6-dichloro-2-fluorophenyl, 1-acetyl-3,6-dichloro-2-fluorobenzene | |
CC(=O)C1=C(C=CC(=C1F)Cl)Cl | |
207.025 | |
207.03 | |
3',6'-Dichloro-2'-fluoroacetophenone |
C8H5Cl2FO | |
MFCD09025392 | |
KIPVBHCZTSPODV-UHFFFAOYSA-N | |
1-(3,6-dichloro-2-fluorophenyl)ethanone | |
46737559 | |
97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures