Learn More
3,6-Dichloropyridazine, 98%, Thermo Scientific Chemicals
£24.52 - £152.76
Chemical Identifiers
CAS | 141-30-0 |
---|---|
Molecular Formula | C4H2Cl2N2 |
Molecular Weight (g/mol) | 148.974 |
MDL Number | MFCD00006466 |
InChI Key | GUSWJGOYDXFJSI-UHFFFAOYSA-N |
Synonym | pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine |
PubChem CID | 67331 |
IUPAC Name | 3,6-dichloropyridazine |
SMILES | C1=CC(=NN=C1Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482527
|
Thermo Scientific Alfa Aesar
A14795.14 |
25 g |
£24.52
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492527
|
Thermo Scientific Alfa Aesar
A14795.22 |
100 g |
£45.12
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402537
|
Thermo Scientific Alfa Aesar
A14795.36 |
500 g |
£152.76
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
141-30-0 | |
148.974 | |
GUSWJGOYDXFJSI-UHFFFAOYSA-N | |
67331 | |
C1=CC(=NN=C1Cl)Cl |
C4H2Cl2N2 | |
MFCD00006466 | |
pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine | |
3,6-dichloropyridazine |
Specifications
141-30-0 | |
C4H2Cl2N2 | |
UN2811 | |
pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine | |
GUSWJGOYDXFJSI-UHFFFAOYSA-N | |
3,6-dichloropyridazine | |
67331 | |
98% |
66°C to 70°C | |
MFCD00006466 | |
111151 | |
Soluble in chloroform. Insoluble in water. | |
C1=CC(=NN=C1Cl)Cl | |
148.974 | |
148.98 | |
3,6-Dichloropyridazine |
Safety and Handling
GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H332-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 205-478-6
missing translation for 'rtecsNumber' : UR5886250
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only