missing translation for 'onlineSavingsMsg'
Learn More

3,6-Difluorophthalic acid, 98%, Thermo Scientific Chemicals

Product Code. 15453377
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 651-97-8
Molecular Formula C8H4F2O4
Molecular Weight (g/mol) 202.113
MDL Number MFCD03840499
InChI Key VFLMWMTWWZGXGA-UHFFFAOYSA-N
Synonym 3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro
PubChem CID 643385
IUPAC Name 3,6-difluorophthalic acid
SMILES C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F

Specifications

Melting Point ∼184°C (decomposition)
Quantity 250 mg
Formula Weight 202.11
Percent Purity 98%
Chemical Name or Material 3,6-Difluorophthalic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'