missing translation for 'onlineSavingsMsg'
Learn More

3,6-Difluorophthalic anhydride, 97%, Thermo Scientific Chemicals

Product Code. 15453427
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-40-4
Molecular Formula C8H2F2O3
Molecular Weight (g/mol) 184.098
MDL Number MFCD00134537
InChI Key AVLRPSLTCCWJKC-UHFFFAOYSA-N
Synonym 3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride
PubChem CID 513918
IUPAC Name 4,7-difluoro-2-benzofuran-1,3-dione
SMILES C1=CC(=C2C(=C1F)C(=O)OC2=O)F

Specifications

Melting Point 218°C to 221°C
Quantity 500 mg
Sensitivity Moisture sensitive
Formula Weight 184.1
Percent Purity 97%
Chemical Name or Material 3,6-Difluorophthalic anhydride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'