missing translation for 'onlineSavingsMsg'
Learn More

3,6-Difluoropyridine-2-carboxylic acid, Thermo Scientific™

Product Code. 10007889
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 851386-45-3
Packaging Amber Glass Bottle
Molecular Formula C6H3F2NO2
Quantity 1 g
Synonym 3,6-difluoropicolinic acid, 3,6-difluoro pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3,6-difluoro, 3,6-difluoro-2-pyridinecarboxylic acid, pubchem11892, 3,6-difluoropyridine-2-carboxylic acid, 3,6-difluoro-pyridine-2-carboxylic acid, 3,6-difluoro-2-pyridinecarboxylic acid;3,6-difluoropicolinic acid
InChI Key FSXFZWBQCAGFHG-UHFFFAOYSA-N
SMILES C1=CC(=NC(=C1F)C(=O)O)F
IUPAC Name 3,6-difluoropyridine-2-carboxylic acid
Molecular Weight (g/mol) 159.092
PubChem CID 11286608
Formula Weight 159.09
Percent Purity 97%
Chemical Name or Material 3, 6-Difluoropyridine-2-carboxylic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.