missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 63°C to 67°C |
Color | White |
Quantity | 5 g |
Beilstein | 8548328 |
Sensitivity | Moisture sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 210.08 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'