missing translation for 'onlineSavingsMsg'
Learn More

3-(9-Anthryl)propionic acid, 96%, Thermo Scientific Chemicals

Product Code. 15427117
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41034-83-7
Molecular Formula C17H14O2
Molecular Weight (g/mol) 250.30
MDL Number MFCD00068641
InChI Key BDGMYCZUEIGHJH-UHFFFAOYSA-N
Synonym 9-anthracenepropionic acid, 3-anthracen-9-yl propanoic acid, 3-anthracen-9-yl-propionic acid, 9-anthracenepropanoic acid, 3-9-anthryl propanoic acid, 9apa, 9-anthracenepropanoicacid, 9-an-thracenepropionic acid, 3-9-anthryl-propionic acid, 3-anthracen-9-ylpropionic acid
PubChem CID 170457
IUPAC Name 3-anthracen-9-ylpropanoic acid
SMILES OC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 193°C to 197°C
Quantity 250 mg
Formula Weight 250.3
Percent Purity 96%
Chemical Name or Material 3-(9-Anthryl)propionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'