missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11432638
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-48-4
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00013983
InChI Key RGDPZMQZWZMONQ-UHFFFAOYSA-N
Synonym 3-acetylamino benzoic acid, benzoic acid, 3-acetylamino, 3-acetylaminobenzoic acid, m-acetylaminobenzoic acid, benzoic acid, m-acetamido, n-acetyl-m-aminobenzoic acid, 3-acetylamino-benzoic acid, 3-acetylamino-benzoicaci, 3-acetamidobenzoicacid, 3'-carboxyacetanilide
PubChem CID 48847
IUPAC Name 3-acetamidobenzoic acid
SMILES CC(=O)NC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 249°C to 252°C
Quantity 500 g
Beilstein 2210867
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 3-Acetamidobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'