missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidophenol, 98%, Thermo Scientific Chemicals

Product Code. 11445103
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-42-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002263
InChI Key QLNWXBAGRTUKKI-UHFFFAOYSA-N
Synonym 3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid
PubChem CID 12124
ChEBI CHEBI:76987
IUPAC Name N-(3-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)O

Specifications

Melting Point 146°C to 149°C
Quantity 500 g
Beilstein 907998
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 3-Acetamidophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'