missing translation for 'onlineSavingsMsg'
Learn More

3-Acetoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 11333057
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Acetoxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions. This material may darken on storage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34231-78-2
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00016603
InChI Key GVUMZPWBUAGJBP-UHFFFAOYSA-N
Synonym 3-acetoxybenzaldehyde, benzaldehyde, 3-acetyloxy, acetoxybenzaldehyde, acetic acid 3-formylphenyl ester, m-acetoxybenzaldehyde, m-ace-toxybenzaldehyde, meta-acetoxybenzaldehyde, pubchem13227, 3-formylphenyl acetate
PubChem CID 118618
IUPAC Name (3-formylphenyl) acetate
SMILES CC(=O)OC1=CC=CC(=C1)C=O

Specifications

Density 1.17
Boiling Point 138°C (20 mmHg)
Refractive Index 1.533
Quantity 5 g
Beilstein 1941499
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 3-Acetoxybenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'