missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,5-dimethylthiophene, 99%, Thermo Scientific Chemicals

Product Code. 11493567
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in the synthesis of heterocyclic ketimines, 3-acetyl-2,5-dimethylthiophene thiosemicarbazone and 3-acetyl-2,5- dimethylthiophene semicarbazone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of heterocyclic ketimines, 3-acetyl-2,5-dimethylthiophene thiosemicarbazone and 3-acetyl-2,5- dimethylthiophene semicarbazone.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-10-1
MDL Number MFCD00009763

Specifications

Density 1.086
Boiling Point 224°C
Flash Point 98°C (208°F)
Molecular Formula C9H22O3Si
Refractive Index 1.5455
Quantity 10 g
Beilstein 112095
Synonym propyltriethoxysilane, triethoxy propyl silane, n-propyltriethoxysilane, silane, triethoxypropyl, unii-f17otx9fg5, f17otx9fg5, dynasylan pteo, triethoxypropyl-silan, triethoxypropyl-silane, triethoxy-n-propylsilane
Solubility Information Soluble in alcohol. Insoluble in water.
InChI Key NBXZNTLFQLUFES-UHFFFAOYSA-N
SMILES CCC[Si](OCC)(OCC)OCC
IUPAC Name triethoxy(propyl)silane
Molecular Weight (g/mol) 206.36
PubChem CID 75691
Formula Weight 154.23
Percent Purity 99%
Chemical Name or Material 3-Acetyl-2,5-dimethylthiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'