missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2-methyl-5-phenylthiophene, 98%, Thermo Scientific Chemicals

Product Code. 11313147
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40932-63-6
Molecular Formula C13H12OS
Molecular Weight (g/mol) 216.30
MDL Number MFCD00151790
InChI Key GREAZYFTAJMZFD-UHFFFAOYSA-N
Synonym 3-acetyl-2-methyl-5-phenylthiophene, 1-2-methyl-5-phenylthiophen-3-yl ethanone, 1-2-methyl-5-phenylthien-3-yl ethanone, ethanone, 1-2-methyl-5-phenyl-3-thienyl, acmc-20anao, 3-acetyl-2-methyl-5-phenylthiohene, 1-2-methyl-5-phenyl-3-thienyl ethanone, 1-2-methyl-5-phenyl-3-thiophenyl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl-ethanone
PubChem CID 2728765
IUPAC Name 1-(2-methyl-5-phenylthiophen-3-yl)ethanone
SMILES CC(=O)C1=C(C)SC(=C1)C1=CC=CC=C1

Specifications

Melting Point 67°C to 69°C
Quantity 5 g
Beilstein 8481276
Formula Weight 216.3
Percent Purity 98%
Chemical Name or Material 3-Acetyl-2-methyl-5-phenylthiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'