missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylindole, 98%, Thermo Scientific Chemicals

Product Code. 11403327
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 703-80-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00005626
InChI Key VUIMBZIZZFSQEE-UHFFFAOYSA-N
Synonym 3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
PubChem CID 12802
IUPAC Name 1-(1H-indol-3-yl)ethanone
SMILES CC(=O)C1=CNC2=CC=CC=C21

Specifications

Melting Point 186°C to 192°C
Quantity 100 g
Beilstein 122579
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 3-Acetylindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'