missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylthiophene, 98%, Thermo Scientific Chemicals

Product Code. 10502981
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1468-83-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00005468
InChI Key RNIDWJDZNNVFDY-UHFFFAOYSA-N
Synonym 3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone
PubChem CID 15116
IUPAC Name 1-thiophen-3-ylethanone
SMILES CC(=O)C1=CSC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 57°C to 61°C
Color Beige to Brown
Boiling Point 208°C to 210°C (748.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C4H3S)COCH3
Quantity 10 g
Beilstein 17,III,452
Formula Weight 126.18
Percent Purity 98%
Physical Form Adhering Crystals
Chemical Name or Material 3-Acetylthiophene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Acetylthiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'