missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-1,2,4-triazole-5-carboxylic acid hydrate, 98%, Thermo Scientific™
Click to view available options
Quantity:
50 g
250 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Amino-1,2,4-triazole-5-carboxylic acid hydrate (AmTAZAc) has been used in the synthesis of new three-dimensional metal-organic framework, [ZnF(AmTAZ)] solvents.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
3-Amino-1,2,4-triazole-5-carboxylic acid hydrate (AmTAZAc) has been used in the synthesis of new three-dimensional metal-organic framework, [ZnF(AmTAZ)] solvents.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼182°C (decomposition) |
Quantity | 250 g |
Beilstein | 124734 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 128.09 (Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | 3-Amino-1,2,4-triazole-5-carboxylic acid hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'